Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl N,N,N',N'-Tetramethylphosphorodiamidate, TCI America™
SDP

Catalog No. T21975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T21975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T21975G Supplier TCI America Supplier No. T21975G

missing translation for 'validMainframeMsg'

(ca. 20% in Benzene)

Chemical Identifiers

CAS 143520-19-8
Molecular Formula C38H47N2O7P
Molecular Weight (g/mol) 674.775
MDL Number MFCD06797176
InChI Key RTYAYXUZHPWJOK-JTDOPDNRSA-N
PubChem CID 11115071
IUPAC Name N-[dimethylamino-[(2R,3R,4S,5R,6R)-3,4,5-tris(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-yl]oxyphosphoryl]-N-methylmethanamine
SMILES CN(C)P(=O)(N(C)C)OC1C(C(C(C(O1)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Color Colorless
Quantity 5 g
UN Number 1114
Formula Weight 674.77
Physical Form Liquid
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl N,N,N′,N′-Tetramethylphosphorodiamidate (c
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.