Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetrafluorobenzoyl chloride, 98%, Thermo Scientific™

Catalog No. AAL2020606
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2020606 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2020606 Supplier Thermo Scientific Chemicals Supplier No. L2020606

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123016-51-3
Boiling Point 94°C to 97°C (61mmHg)
Molecular Formula C7HClF4O
Quantity 5g
MDL Number MFCD03001171
UN Number UN3265
Beilstein 6398940
Synonym acmc-20aoip, 2,3,4,6-tetrafluorobenzoylchloride, benzoyl chloride, 2,3,4,6-tetrafluoro-9ci
InChI Key UYKQDVFTGGRFEQ-UHFFFAOYSA-N
SMILES FC1=CC(F)=C(C(Cl)=O)C(F)=C1F
IUPAC Name 2,3,4,6-tetrafluorobenzoyl chloride
Molecular Weight (g/mol) 212.53
PubChem CID 2783276
Formula Weight 212.53
Percent Purity 98%
Chemical Name or Material 2,3,4,6-Tetrafluorobenzoyl chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.