Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,4-Dimethoxyphenyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. D410725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D410725G 25 g
D41075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D410725G Supplier TCI America Supplier No. D410725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7417-21-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00002894
InChI Key SRQAJMUHZROVHW-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene
PubChem CID 81911
IUPAC Name 2-(3,4-dimethoxyphenyl)ethan-1-ol
SMILES COC1=CC=C(CCO)C=C1OC

Specifications

Melting Point 46°C
Boiling Point 174°C
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.