Learn More
Medchemexpress LLC 2,3,4-tri-O-acetyl-D-glucuronide methyl ester | 3082-95-9 | 99.7% | 334.28 g/mol | C13H18O10 | 100 MG

Supplier: Medchemexpress LLC HYW039885100MG
2,3,4-Tri-O-acetyl-D-glucuronide methyl ester is an acetyl-protected methyl ester of D-glucuronic acid used as a synthetic intermediate and analytical reference in carbohydrate and organic synthesis. It is supplied as a light-yellow solid with accompanying analytical documentation for research use.
- High purity 99.7% for analytical and synthetic applications.
- Molecular weight 334.28 g/mol.
- Soluble in common organic solvents such as DMSO and methanol.
- Supplied with COA, RP-HPLC, MS, and NMR documentation.
- Suitable as a synthetic intermediate and reference standard for carbohydrate chemistry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.