Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3',4'-Trihydroxyacetophenone, 98%

Catalog No. AAA1436414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1436414 25 g
AAA1436406 5 g
AAA1436422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1436414 Supplier Thermo Scientific Chemicals Supplier No. A1436414
Add to Cart

CAS: 528-21-2 | C8H8O4 | 168.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-21-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002193
InChI Key XIROXSOOOAZHLL-UHFFFAOYSA-N
Synonym gallacetophenone, 1-2,3,4-trihydroxyphenyl ethanone, 2',3',4'-trihydroxyacetophenone, galloacetophenone, alizarin yellow c, alizarine yellow c, 2,3,4-trihydroxyacetophenone, ethanone, 1-2,3,4-trihydroxyphenyl, 4-acetylpyrogallol, acetophenone, 2',3',4'-trihydroxy
PubChem CID 10706
IUPAC Name 1-(2,3,4-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)O)O)O

Specifications

Melting Point 169°C to 172°C
Quantity 25 g
Merck Index 14,4342
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 2',3',4'-Trihydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-430-2
RTECSNumber AN0527000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.