Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoro-4-aminobenzotrifluoride 97.0+%, TCI America™
SDP

Catalog No. T15291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15291G 1 g
T15295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15291G Supplier TCI America Supplier No. T15291G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 651-83-2
Molecular Formula C7H2F7N
Molecular Weight (g/mol) 233.089
MDL Number MFCD00091518
InChI Key FJOACTZFMHZHSC-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-4-trifluoromethyl aniline, 2,3,5,6-tetrafluoro-4-aminobenzotrifluoride, 4-amino-heptafluorotoluene, 4-amino-2,3,5,6-tetrafluorobenzotrifluoride, 4-aminotetrafluorobenzotrifluoride, 4-aminoheptafluorotoluene, 2,3,5,6-tetrafluoro-4-trifluoromethyl phenylamine, benzenamine, 2,3,5,6-tetrafluoro-4-trifluoromethyl, pubchem2817, yu-wasa auxiliary
PubChem CID 616257
IUPAC Name 2,3,5,6-tetrafluoro-4-(trifluoromethyl)aniline
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 186°C
Quantity 1 g
Formula Weight 233.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluoro-4-aminobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.