Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Tribromothiophene 98.0+%, TCI America™
SDP

Catalog No. T0956250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0956250G 250 g
T095625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0956250G Supplier TCI America Supplier No. T0956250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3141-24-0
Molecular Formula C4HBr3S
Molecular Weight (g/mol) 320.82
MDL Number MFCD00014521
InChI Key SKDNDSLDRLEELJ-UHFFFAOYSA-N
Synonym thiophene, 2,3,5-tribromo, pubchem5608, 2,5-tribromothiophene, acmc-209hmg, thiophene,3,5-tribromo, 2,3,5-tribromo thiophene, ksc225k9l, 2,3,5-tribromothiophene
PubChem CID 76592
IUPAC Name 2,3,5-tribromothiophene
SMILES BrC1=CC(Br)=C(Br)S1

Specifications

Color Yellow
Quantity 250 g
Formula Weight 320.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,5-Tribromothiophene
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.