Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trifluoropyridine 98.0+%, TCI America™
SDP

Catalog No. T27785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27785G 5 g
T277825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27785G Supplier TCI America Supplier No. T27785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76469-41-5
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001162
InChI Key JKVOXNTXYMXDHN-UHFFFAOYSA-N
Synonym pyridine, 2,3,5-trifluoro, pubchem8020, 2,3,5-trifluoro-pyridine, ksc494e2b, 2,3,5-tris fluoranyl pyridine, 2,3,5-trifluoropyridine 1g
PubChem CID 2783287
IUPAC Name 2,3,5-trifluoropyridine
SMILES C1=C(C=NC(=C1F)F)F

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 1993
Formula Weight 133.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trifluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.