Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2,3,5-Triphenyltetrazolium Chloride

Catalog No. MP219989225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
50 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219989225 25 g
MP219989201 1 g
MP219989210 10 g
MP219989250 50 g
MP219989280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP219989225 Supplier MP Biomedicals™ Supplier No. 0219989225
Add to Cart
Add to Cart

2,3,5-Triphenyltetrazolium Chloride is a redox indicator.

  • In the TTC assay (also known as TTC test or tetrazolium test), TTC is used to differentiate between metabolically active and inactive tissues.
  • The white compound is enzymatically reduced to red TPF (1,3,5-triphenylformazan) in living tissues due to the activity of various dehydrogenases (enzymes important in oxidation of organic compounds and thus cellular metabolism), while it remains as white TTC in areas of necrosis since these enzymes have been either denatured or degraded.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
Storage Requirements Store at +4°C, protect from light.
Product Type Triphenyltetrazolium Chloride

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.