Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Triphenyltetrazolium Chloride, Reagents

Catalog No. 01336903
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-903 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-903 Supplier Reagents Holdings Llc Supplier No. C23427005C2
Add to Cart
Add to Cart

2,3,5-Triphenyltetrazolium Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 298-96-4
Molecular Formula C19H15ClN4
Molecular Weight (g/mol) 334.81
InChI Key PKDBCJSWQUOKDO-UHFFFAOYSA-M
IUPAC Name triphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White to Yellow
Packaging Glass Bottle
Quantity 5 g
Specific Gravity 1
Grade Reagent
Physical Form Solid
Chemical Name or Material 2,3,5-Triphenyltetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.