Learn More
Sigma Organic Chemistry 2- 3-Aminophenyl -4-phe | 1G | MFCD06739766
Supplier: Sigma Organic Chemistry BOG001861G

2- 3-Aminophenyl -4-phe , 1G
About this Item:
MDL #: MFCD06739766
Molecular Weight: 266.36
UNSPSC Code: 12352200
Chemical Formula: C16H14N2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.