Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Benzanthracene, 98%

Catalog No. AC105260050
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105260050 5 g Glass bottle
AC105265000 500 mg Glass bottle
AC105260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC105260050 Supplier Thermo Scientific Chemicals Supplier No. 105260050
Add to Cart

CAS: 92-24-0 | C18H12 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

Specifications

Melting Point >300°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 05,IV,2545
Merck Index 15,6454
Solubility Information Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene
Formula Weight 228.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Benzanthracene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 202-138-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.