Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Benzofuran 99.0+%, TCI America™
SDP

Catalog No. B006025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006025G 25 g
B00605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B006025G Supplier TCI America Supplier No. B006025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12

Specifications

Boiling Point 175°C
Quantity 25 g
UN Number 1993
Formula Weight 118.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Benzofuran
RTECSNumber DF6423800
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.