Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide 97.0+%, TCI America™
SDP

Catalog No. B1535100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1535100MG 100 mg
B15351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1535100MG Supplier TCI America Supplier No. B1535100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 220191-36-6
Molecular Formula C18H19NO2S2
Molecular Weight (g/mol) 345.48
MDL Number MFCD00142790
InChI Key OHZCQTZIDIVCPI-UHFFFAOYSA-N
PubChem CID 22023748
IUPAC Name 3,4-bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
SMILES CC1=C(C)C(=C(C)S1)C1=C(C(=O)NC1=O)C1=C(C)SC(C)=C1C

Specifications

Melting Point 181°C
Quantity 100 mg
Formula Weight 345.48
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(2,4,5-trimethyl-3-thienyl)maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.