Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromophenyl)ethyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B351725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B351725G 25 g
B35175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B351725G Supplier TCI America Supplier No. B351725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28229-69-8
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00191857
InChI Key PTTFLKHCSZSFOL-UHFFFAOYSA-N
Synonym 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
PubChem CID 2734090
IUPAC Name 2-(3-bromophenyl)ethan-1-ol
SMILES OCCC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromophenyl)ethyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.