Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanedithiol 98.0+%, TCI America™
SDP

Catalog No. B288825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B288825G 25 g
B28885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B288825G Supplier TCI America Supplier No. B288825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4532-64-3
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.244
MDL Number MFCD00010024
InChI Key TWWSEEHCVDRRRI-UHFFFAOYSA-N
Synonym 2,3-Dimercaptobutane
PubChem CID 548353
IUPAC Name butane-2,3-dithiol
SMILES CC(C(C)S)S

Specifications

Color Yellow
Boiling Point 87°C
Quantity 25 g
UN Number 3336
Formula Weight 122.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Butanedithiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.