Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chloropropyl)-1,3-dioxolane 97.0+%, TCI America™
SDP

Catalog No. C30335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30335G Supplier TCI America Supplier No. C30335G

Chemical Identifiers

CAS 16686-11-6
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00043135
InChI Key ZBPUNVFDQXYNDY-UHFFFAOYSA-N
Synonym 4-Chlorobutyraldehyde Ethylene Acetal
PubChem CID 552072
IUPAC Name 2-(3-chloropropyl)-1,3-dioxolane
SMILES ClCCCC1OCCO1

Specifications

Boiling Point 53°C
Quantity 5 g
Formula Weight 150.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Chloropropyl)-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.