Learn More
Sigma Organic Chemistry 2- 3-Cyanophenyl benzim | 1G | MFCD11218395
Supplier: Sigma Organic Chemistry BOG001931G

2- 3-Cyanophenyl benzim , 1G
About this Item:
MDL #: MFCD11218395
Molecular Weight: 219.24
UNSPSC Code: 12352200
Chemical Formula: C14H9N3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.