Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diamino-5-bromopyridine 98.0+%, TCI America™
SDP

Catalog No. D291625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D291625G 25 g
D29165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D291625G Supplier TCI America Supplier No. D291625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38875-53-5
Molecular Formula C5H7BrN3
Molecular Weight (g/mol) 189.04
MDL Number MFCD00460094
InChI Key YRGMYJUKFJPNPD-UHFFFAOYSA-O
Synonym 2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine
PubChem CID 691156
IUPAC Name 2,3-diamino-5-bromopyridin-1-ium
SMILES NC1=CC(Br)=C[NH+]=C1N

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 188.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diamino-5-bromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.