Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropanol 96%, Thermo Scientific™

Catalog No. AC112890050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC112890050 5 g
AC112891000 100 g
AC112895000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112890050 Supplier Thermo Scientific Chemicals Supplier No. 112890050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-13-9
Molecular Formula C3H6Br2O
Molecular Weight (g/mol) 217.89
MDL Number MFCD00004699
InChI Key QWVCIORZLNBIIC-UHFFFAOYNA-N
Synonym 2,3-dibromo-1-propanol, 2,3-dibromopropanol, 1-propanol, 2,3-dibromo, dibromopropanol, brominex 257, 2,3-dibromopropyl alcohol, 1,2-dibromopropan-3-ol, usaf do-42, dbp flame retardant, beta-dibromohydrin
PubChem CID 7281
ChEBI CHEBI:19310
IUPAC Name 2,3-dibromopropan-1-ol
SMILES OCC(Br)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 2.12
Infrared Spectrum Authentic
Refractive Index 1.5570 to 1.5590 (20°C, 589nm)
Linear Formula BrCH2CH(Br)CH2OH
Quantity 5 g
Beilstein 01, 357
Formula Weight 217.89
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 2, 3-Dibromopropanol, 96%
Show More Show Less
EINECSNumber 202-480-9
RTECSNumber UB0175000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.