Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Dibromopropene

Catalog No. 10600325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-600-325G 25 g
10-600-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-600-325G Supplier Sigma Aldrich Supplier No. 10600325G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,3-dibromopropene is an intermediate formed during the hydrolysis of nematocide:1,2-dibromo-3-chloropropane.2,3-dibromopropene was used to test 2- and 3-carbon halogenated hydrocarbons for mutagenicity for salmonella typhimurium strain ta 100
  • It was used in the synthesis of n(boc)-L-(2-bromoallyl)-glycine

Specifications

CAS 513-31-5
Density 2.045 g/mL (at 25°C)
Boiling Point 42°C to 44°C (17 mmHg)
Molecular Formula C3H4Br2
Linear Formula CH2BrCBr=CH2
Refractive Index n20/D 1.544 (literature)
MDL Number MFCD00000211
Quantity 25 g
Molecular Weight (g/mol) 199.87
Percent Purity 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.