Learn More
2,3-Dichloro-1,4-naphthoquinone, 98%
Description
Use in textiles, as organic catalyst, as chemical intermediate, as additive in dye binders. Also used as an important catalyst to produce 3,3-dichlorobenzene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Use as seed disinfectant, textiles, as organic catalyst, as chemical intermediate, as additive in dye binders, as antifouling agent. Also used to control blue-green algae in ponds, lakes, and swimming pools. Also used as an important catalyst to produce 3,3-Dichloro benzine.
Solubility
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 193°C to 196°C (sublimation) |
| Boiling Point | 275°C (2 mmHg) |
| Quantity | 100 g |
| UN Number | UN2811 |
| Beilstein | 1073511 |
| Merck Index | 14,3045 |
| Solubility Information | Soluble in water (0.008g/L). Soluble in hot toluene almost transparency. |
| IUPAC Name | 2,3-dichloronaphthalene-1,4-dione |
| Formula Weight | 227.05 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H500 |
| Hazard Statement | GHS H Statement H302-H315-H319 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 204-210-5 |
| RTECSNumber | QL7525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.