Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. D44161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44161G 1 g
D44165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44161G Supplier TCI America Supplier No. D44161G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22353-40-8
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD03840432
InChI Key XLPDVAVQKGDHNO-UHFFFAOYSA-N
PubChem CID 2763106
IUPAC Name 2,3-dichloro-5-nitropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)[N+](=O)[O-]

Specifications

Melting Point 53°C
Color Yellow
Boiling Point 256°C
Quantity 1 g
Formula Weight 192.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.