Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloroanisole 97.0+%, TCI America™
SDP

Catalog No. D230625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D230625G 25 g
D2306500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D230625G Supplier TCI America Supplier No. D230625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1984-59-4
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00000545
InChI Key HFEASCCDHUVYKU-UHFFFAOYSA-N
Synonym 2,3-dichloroanisole, benzene, 1,2-dichloro-3-methoxy, 2,3-dichloromethoxybenzene, 2,3-dichloranisol, dichloroanisole 2,3-, 1,2-dichloro-3-methoxy-benzene, dichloroanisol, 2,3-dichloro-anisol, pubchem3680, 2,3-dichloro anisole
PubChem CID 16126
IUPAC Name 1,2-dichloro-3-methoxybenzene
SMILES COC1=CC=CC(Cl)=C1Cl

Specifications

Quantity 25 g
Formula Weight 177.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3-Dichloroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.