Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzenethiol 95.0+%, TCI America™
SDP

Catalog No. D190325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D190325G 25 g
D19035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D190325G Supplier TCI America Supplier No. D190325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17231-95-7
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041420
InChI Key QGRKONUHHGBHRB-UHFFFAOYSA-N
Synonym 2,3-dichlorothiophenol, 2,3-dichlorobenzene-1-thiol, dichlorothiophenol, benzenethiol, dichloro, 2,3-dichloro thiophenol, pubchem6796, 2,3-dichiorothiophenol, 2,3-dichloro-thiophenol, acmc-1cf9p, 2,3-dichlorobenzene thiol
PubChem CID 140202
IUPAC Name 2,3-dichlorobenzenethiol
SMILES C1=CC(=C(C(=C1)Cl)Cl)S

Specifications

Melting Point 59°C
Boiling Point 100°C
Quantity 25 g
Formula Weight 179.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichlorobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.