Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloropyrazine 98.0+%, TCI America™
SDP

Catalog No. D306125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D306125G 25 g
D30615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D306125G Supplier TCI America Supplier No. D306125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4858-85-9
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00040964
InChI Key MLCNOCRGSBCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
PubChem CID 78575
IUPAC Name 2,3-dichloropyrazine
SMILES ClC1=NC=CN=C1Cl

Specifications

Color White-Yellow
Boiling Point 107°C
Quantity 25 g
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3-Dichloropyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.