Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluoro-6-nitroaniline, 97%, Thermo Scientific Chemicals

Catalog No. AAH6325122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6325122 100 g
AAH6325114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6325122 Supplier Thermo Scientific Chemicals Supplier No. H6325122

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211693-73-1
Molecular Formula C6H4F2N2O2
Molecular Weight (g/mol) 174.11
MDL Number MFCD00233313
InChI Key DIPGYZSCGXBTEU-UHFFFAOYSA-N
Synonym 2-amino-3,4-difluoronitrobenzene, 2,3-difluoro-6-nitrophenylamine, 2,3-difluoro-6-nitrobenzenamine, benzenamine, 2,3-difluoro-6-nitro, acmc-1byjz, 5,6-difluoro-2-nitroaniline, ksc495e7r, 2,3-difluoro-6-nitro-aniline, buttpark 15\01-97, 2,3-difluoro-6-nitro-phenylamine
PubChem CID 2737028
IUPAC Name 2,3-difluoro-6-nitroaniline
SMILES NC1=C(C=CC(F)=C1F)[N+]([O-])=O

Specifications

Quantity 100 g
UN Number UN2811
Formula Weight 174.11
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-6-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.