Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D205825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D205825G 25 g
D20585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. D205825G Supplier TCI America Supplier No. D205825G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 4519-39-5
Molecular Formula C7H3F2O2
Molecular Weight (g/mol) 157.10
MDL Number MFCD00010267
InChI Key JLZVIWSFUPLSOR-UHFFFAOYSA-M
Synonym 2,3-difluoro-benzoic acid, benzoic acid, 2,3-difluoro, difluorobenzoic acid, difluorobenzoic acid;, pubchem1379, 2,3 difluorobenzoic acid, acmc-209k0x, 2,3-difluoro benzoic acid, 2,3-difluorobenzoic acid;, ksc235o6t
PubChem CID 370590
IUPAC Name 2,3-difluorobenzoate
SMILES [O-]C(=O)C1=CC=CC(F)=C1F

Specifications

Melting Point 164°C
Color White
Quantity 25 g
Formula Weight 158.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Difluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.