Learn More
2,3-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| CAS | 121219-16-7 |
| Color | White-Yellow |
| Molecular Formula | C6H5BF2O2 |
| MDL Number | MFCD01863170 |
| Quantity | 25 g |
| Synonym | 2,3-difluorobenzeneboronic acid, 2,3-difluorophenyl boronic acid, 2,3-difluorophenyl boranediol, 2,3-difluorophenylboronicacid, 2,3-difluoro phenylboric acid |
| InChI Key | SZYXKFKWFYUOGZ-UHFFFAOYSA-N |
| SMILES | B(C1=C(C(=CC=C1)F)F)(O)O |
| IUPAC Name | (2,3-difluorophenyl)boronic acid |
| Molecular Weight (g/mol) | 157.911 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.