Learn More
Sigma Organic Chemistry 2 3-diHYD-1 4-benzodiox | 100mg | MFCD04115371
Supplier: Sigma Organic Chemistry CDS013915100MG

2 3-diHYD-1 4-benzodiox , 100mg
About this Item:
MDL #: MFCD04115371
Molecular Weight: 201.65
UNSPSC Code: 12352200
Chemical Formula: C9H12ClNO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.