Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-1-benzofuran-5-yl isocyanate, 97%, Maybridge™

Catalog No. CC00806DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00806DA 1 g
CC00806CB 250 mg
CC00806DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC00806DA Supplier Thermo Scientific Chemicals Supplier No. CC00806DA
Add to Cart

Chemical Identifiers

CAS 215162-92-8
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD02677718
InChI Key WAIKTAFMGLVRJC-UHFFFAOYSA-N
Synonym 2,3-dihydro-1-benzofuran-5-yl isocyanate, 5-isocyanato-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-ylisocyanate, 2,3-dihydrobenzo b furan-5-isocyanate, benzofuran,2,3-dihydro-5-isocyanato, 2,3-dihydrobenzofuran-5-yl isocyanate, 2,3-dihydro-5-isocyanatobenzo b furan, 5-isocyanato-2,3-dihydrobenzo b furan, 2,3-dihydro-5-isocyanato-1-benzofuran, 2,3-dihydro-1-benzofuran-5-yl-isocyanate
PubChem CID 2776156
IUPAC Name 5-isocyanato-2,3-dihydro-1-benzofuran
SMILES C1COC2=C1C=C(C=C2)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1-benzofuran-5-yl isocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.