Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran 98.0+%, TCI America™
SDP

Catalog No. D215425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D215425G 25 g
D2154100G 100 g
D2154500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D215425G Supplier TCI America Supplier No. D215425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1563-38-8
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00075382
InChI Key WJGPNUBJBMCRQH-UHFFFAOYSA-N
Synonym 2,2-Dimethyl-7-hydroxycoumaran
PubChem CID 15278
ChEBI CHEBI:38474
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-ol
SMILES CC1(CC2=C(O1)C(=CC=C2)O)C

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dihydro-2,2-dimethyl-7-hydroxybenzofuran
EINECSNumber (5)-5541
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.