Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-Dihydroxy-4'-methoxyacetophenone Hydrate 98.0+%, TCI America™
SDP

Catalog No. D218725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D218725G 25 g
D21875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D218725G Supplier TCI America Supplier No. D218725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 708-53-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00180772
InChI Key VCONERRCKOKCHE-UHFFFAOYSA-N
PubChem CID 12820
IUPAC Name 1-(2,3-dihydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)OC)O)O

Specifications

Melting Point 133°C
Quantity 25 g
Synonym Gallacetophenone-4′-methyl Ether
Formula Weight 182.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Dihydroxy-4′-methoxyacetophenone Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.