Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydroxybenzoic acid, 99%

Catalog No. p-112057
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155170050 5 g Glass bottle
AC155170250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155170050 Supplier Thermo Scientific Chemicals Supplier No. 155170050
Only null left
Add to Cart

CAS: 303-38-8 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-38-8
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002446
InChI Key GLDQAMYCGOIJDV-UHFFFAOYSA-N
Synonym pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
PubChem CID 19
ChEBI CHEBI:18026
IUPAC Name 2,3-dihydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color Beige to Brown or Gray-Pink
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2H
Quantity 5 g
Beilstein 10, 375
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dihydroxybenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 206-139-5
RTECSNumber DG8576490
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.