Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzylamine 98.0+%, TCI America™
SDP

Catalog No. D40225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40225G Supplier TCI America Supplier No. D40225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4393-09-3
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.21
MDL Number MFCD00052392
InChI Key LVMPWFJVYMXSNY-UHFFFAOYSA-N
Synonym 2,3-dimethoxybenzylamine, 2,3-dimethoxyphenyl methanamine, 1-2,3-dimethoxyphenyl methanamine, benzenemethanamine, 2,3-dimethoxy, 2,3-dimethoxy-benzylamine, 2,3-dimethoxy benzylamine, pubchem7399, 2,3-dimethoxybenzyl amine, acmc-2097fc
PubChem CID 78106
IUPAC Name (2,3-dimethoxyphenyl)methanamine
SMILES COC1=CC=CC(=C1OC)CN

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
UN Number 2735
Formula Weight 167.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.