Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. D428925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D428925G 25 g
D42895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D428925G Supplier TCI America Supplier No. D428925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 169051-76-7
Molecular Formula C10H15F6N3O4S2
Molecular Weight (g/mol) 419.357
MDL Number MFCD03788922
InChI Key XOZHIVUWCICHSQ-UHFFFAOYSA-N
Synonym 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide
PubChem CID 11750474
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1,2-dimethyl-3-propylimidazol-1-ium
SMILES CCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 8°C
Color Yellow
Quantity 25 g
Formula Weight 419.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethyl-1-propylimidazolium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.