Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylanthraquinone 95.0+%, TCI America™
SDP

Catalog No. D29801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29801G 1 g
D29805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29801G Supplier TCI America Supplier No. D29801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6531-35-7
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
InChI Key KIJPZYXCIHZVGP-UHFFFAOYSA-N
PubChem CID 81019
IUPAC Name 2,3-dimethylanthracene-9,10-dione
SMILES CC1=C(C=C2C(=C1)C(=O)C3=CC=CC=C3C2=O)C

Specifications

Melting Point 210°C
Color Yellow
Quantity 1 g
Formula Weight 236.27
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.