Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D222125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D222125G 25 g
D22215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D222125G Supplier TCI America Supplier No. D222125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 603-79-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002479
InChI Key RIZUCYSQUWMQLX-UHFFFAOYSA-N
Synonym hemellitic acid, benzoic acid, 2,3-dimethyl, 2,3-dimethylbenzoicacid, 2,3-dimethyl benzoic acid, 2,3-dimethyl-benzoic acid, unii-7irp8ca267, 2,3-dimethylbenzenecarboxylic acid, vic-o-xylylic acid, methyl m-toluic acid, pubchem2549
PubChem CID 11782
ChEBI CHEBI:64823
IUPAC Name 2,3-dimethylbenzoic acid
SMILES CC1=CC=CC(=C1C)C(=O)O

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 150.18
Percent Purity ≥98.0% (T)
Chemical Name or Material 2,3-Dimethylbenzoic Acid
RTECSNumber DG8734000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.