Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzonitrile 97.0+%, TCI America™
SDP

Catalog No. D429325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D429325G 25 g
D42935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D429325G Supplier TCI America Supplier No. D429325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5724-56-1
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.18
MDL Number MFCD00019744
InChI Key ZEDPQIJYJCPIRM-UHFFFAOYSA-N
Synonym benzonitrile, 2,3-dimethyl, 3-cyano-o-xylene, unii-6s215f38ok, 2,3-dimethylbenzonitril german, 2,3-dimethylbenzenecarbonitrile, 2,3-dimethylbenzonitril, dimethylbenzonitrile, pubchem8125, benzonitrile, dimethyl, medetomidine impurity 11
PubChem CID 79802
IUPAC Name 2,3-dimethylbenzonitrile
SMILES CC1=CC=CC(C#N)=C1C

Specifications

Melting Point 16°C
Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 62°C
Formula Weight 131.18
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2,3-Dimethylbenzonitrile
RTECSNumber DI4357600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.