Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylphenol 98.0+%, TCI America™
SDP

Catalog No. D077325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D077325G 25 g
D0773500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D077325G Supplier TCI America Supplier No. D077325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 526-75-0
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00002227
InChI Key QWBBPBRQALCEIZ-UHFFFAOYSA-N
Synonym 1-hydroxy-2,3-dimethylbenzene, 2,3-xylenol, dimethylphenol, xylenol, phenol, 2,3-dimethyl, o-xylenol, vic-o-xylenol, o-3-xylenol, xyellenol 100, 2,3-dimethyl phenol
PubChem CID 10687
IUPAC Name 2,3-dimethylphenol
SMILES CC1=C(C(=CC=C1)O)C

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 218°C
Quantity 25 g
UN Number 2261
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE5500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.