Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylquinoxaline 98.0+%, TCI America™
SDP

Catalog No. D0795100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0795100G 100 g
D079525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0795100G Supplier TCI America Supplier No. D0795100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2379-55-7
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 130°C
Quantity 100 g
Synonym quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
Formula Weight 158.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylquinoxaline
RTECSNumber VD1960000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.