Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride 97.0+%, TCI America™
SDP

Catalog No. B0325100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0325100MG 100 mg
B03251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0325100MG Supplier TCI America Supplier No. B0325100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38800-20-3
Molecular Formula C17H13ClN4S
Molecular Weight (g/mol) 340.829
MDL Number MFCD00060088
InChI Key RLHVMOZOYHDIGV-UHFFFAOYSA-M
Synonym STC
PubChem CID 6097060
IUPAC Name 2,3-diphenyl-5-thiophen-2-yltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=CC=C3)C4=CC=CS4.[Cl-]

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 340.83
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenyl-5-(2-thienyl)tetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.