Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Naphthalenedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. N052625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N052625G 25 g
N05265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N052625G Supplier TCI America Supplier No. N052625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2169-87-1
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004104
InChI Key KHARCSTZAGNHOT-UHFFFAOYSA-N
Synonym 2,3-naphthalenedicarboxylic acid, 2,3-naphthalenedicarboxylicacid, acmc-1cb2t, ghl.pd_mitscher_leg0.770
PubChem CID 95073
IUPAC Name naphthalene-2,3-dicarboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)C(=O)O

Specifications

Melting Point 246°C
Color White
Quantity 25 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Naphthalenedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.