Learn More
Sigma Organic Chemistry 2- 3-Nitrophenyl -4- 4- | 1G | MFCD28166227
Supplier: Sigma Organic Chemistry BOG001821G

2- 3-Nitrophenyl -4- 4- , 1G
About this Item:
MDL #: MFCD28166227
Molecular Weight: 298.32
UNSPSC Code: 12352200
Chemical Formula: C15H10N2O3S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.