Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Norbornanedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. N075325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N075325G 25 g
N07535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N075325G Supplier TCI America Supplier No. N075325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1724-08-9
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
MDL Number MFCD00520577
InChI Key IVVOCRBADNIWDM-UHFFFAOYNA-N
Synonym Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid
PubChem CID 99835
IUPAC Name bicyclo[2.2.1]heptane-2,3-dicarboxylic acid
SMILES OC(=O)C1C2CCC(C2)C1C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 184.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Norbornanedicarboxylic Acid
RTECSNumber RB7168000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.