Learn More
Medchemexpress LLC 2,3-O-isopropylidene-D-erythronolactone | 25581-41-3 | MFCD00134440 | 99.5% | 158.15 g/mol | C7H10O4 | 1 G
Supplier: Medchemexpress LLC HYW1455831G
2,3-O-Isopropylidene-D-erythronolactone is an isopropylidene-protected erythronolactone used as a building block and reagent in organic synthesis and carbohydrate chemistry. Supplied for research applications, it is commonly used in spiroannulation, stereoselective transformations, and small-molecule synthesis workflows.
- High purity (reported 99.47%).
- Defined chemical identity (CAS 25581-41-3).
- Soluble in DMSO at approximately 10 mM, facilitating solution-phase reactions.
- Useful as a protected carbohydrate-derived building block.
- Available in multiple gram-scale pack sizes for research laboratories.
- Intended for research use only (RUO).