Learn More
Medchemexpress LLC 2,3-O-isopropylidene-D-erythronolactone | 25581-41-3 | MFCD00134440 | 99.5% | 158.15 g·mol⁻¹ | C7H10O4 | 5 G
Supplier: Medchemexpress LLC HYW1455835G
2,3-O-isopropylidene-D-erythronolactone is an isopropylidene-protected derivative of D-erythronolactone supplied as a solid reagent for research and organic synthesis. It acts as a protected diol intermediate used in multi-step syntheses where temporary protection of hydroxyl groups is required.
- Provides temporary protection of vicinal diols during synthesis.
- Serves as a versatile building block for carbohydrate and heterocycle construction.
- High purity suitable for synthetic and research applications.
- Available in multiple package sizes for lab-scale use.
- Stable solid form with defined molecular formula and molecular weight.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.