Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-2,3-O-Isopropylidene-L-threitol 97.0+%, TCI America™
SDP

Catalog No. I03761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03761G 1 g
I03765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03761G Supplier TCI America Supplier No. I03761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50622-09-8
Molecular Formula C7H14O4
Molecular Weight (g/mol) 162.185
MDL Number MFCD00063761
InChI Key INVRLGIKFANLFP-WDSKDSINSA-N
Synonym +-2,3-o-isopropylidene-l-threitol, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl dimethanol, 2,3-o-isopropylidene-l-threitol, 4s,5s-5-hydroxymethyl-2,2-dimethyl-1,3-dioxolan-4-yl methanol, +-2,3,o-isopropylidene-l-threitol, pubchem19000, 2,3-o-isoproylidene-l-threitol, 1,3-dioxolane-4,5-dimethanol, 2,2-dimethyl-, 4s,5s, 2-o,3-o-isopropylidene-l-threitol, oi-2,3-o-isopropylidene-l-threitol
PubChem CID 785242
IUPAC Name [(4S,5S)-5-(hydroxymethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OC(C(O1)CO)CO)C

Specifications

Melting Point 46°C
Color White-Yellow
Quantity 1 g
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (+)-2,3-O-Isopropylidene-L-threitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.