Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Thiophenedicarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. T23881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23881G 1 g
T23885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23881G Supplier TCI America Supplier No. T23881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 932-41-2
Molecular Formula C6H4O2S
Molecular Weight (g/mol) 140.156
MDL Number MFCD00209616
InChI Key WSEJZRIZDQWMKQ-UHFFFAOYSA-N
PubChem CID 573752
IUPAC Name thiophene-2,3-dicarbaldehyde
SMILES C1=CSC(=C1C=O)C=O

Specifications

Melting Point 79°C
Boiling Point 134°C
Quantity 1 g
Formula Weight 140.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Thiophenedicarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.