Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Thiophenedicarboxylic Anhydride 98.0+%, TCI America™
SDP

Catalog No. T33421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33421G 1 g
T33425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33421G Supplier TCI America Supplier No. T33421G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6007-83-6
Molecular Formula C6H2O3S
Molecular Weight (g/mol) 154.139
MDL Number MFCD09832453
InChI Key FIDKFEIEZJGDBE-UHFFFAOYSA-N
PubChem CID 222951
IUPAC Name thieno[2,3-c]furan-4,6-dione
SMILES C1=CSC2=C1C(=O)OC2=O

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Thiophenedicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.